Assay Details
| Gene Symbol: tnfaip2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030216.1 | NP_001025387.1 | 10 - 11 | 1746 | 114 | |
| XM_005160467.4 | XP_005160524.1 | 11 - 12 | 1826 | 114 | ||
More Information
Set Membership: |
|

tnfaip2a
Genomic Map