Assay Details
| Gene Symbol: tnfaip2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030216.1 | NP_001025387.1 | 11 - 12 | 1902 | 131 | |
| XM_005160467.4 | XP_005160524.1 | 12 - 13 | 1982 | 131 | ||
| GenBank mRNA | BC114226.1 | - | 11 - 12 | 1906 | 131 | |
More Information
Set Membership: |
|

tnfaip2a
Genomic Map