Assay Details
Gene Symbol: itgb1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034987.1 | NP_001030159.1 | 56 | 73 | ||
GenBank mRNA | BC162706.1 | - | 153 | 73 | ||
DQ149102.1 | - | 56 | 73 | |||
HQ538840.1 | - | 326 | 73 |
Gene Symbol: LOC110437890 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021466326.1 | XP_021322001.1 | 2 - 3 | 396 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
