Assay Details
| Gene Symbol: itgb1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034987.1 | NP_001030159.1 | 533 | 82 | ||
| GenBank mRNA | BC162706.1 | - | 630 | 82 | ||
| DQ149102.1 | - | 533 | 82 | |||
| HQ538840.1 | - | 803 | 82 | |||
| Gene Symbol: LOC110437890 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021466326.1 | XP_021322001.1 | 5 - 6 | 873 | 82 | |
More Information
Set Membership: |
|

itgb1b
Genomic Map