Assay Details
Gene Symbol: itgb1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034987.1 | NP_001030159.1 | 774 | 111 | ||
GenBank mRNA | BC162706.1 | - | 871 | 111 | ||
DQ149102.1 | - | 774 | 111 | |||
HQ538840.1 | - | 1044 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
