Assay Details
| Gene Symbol: svila | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035261.2 | NP_001030338.2 | 36 - 37 | 5534 | 86 | |
| XM_021480011.1 | XP_021335686.1 | 42 - 43 | 7029 | 86 | ||
| XM_021480012.1 | XP_021335687.1 | 41 - 42 | 6959 | 86 | ||
| XM_021480013.1 | XP_021335688.1 | 41 - 42 | 6953 | 86 | ||
| XM_021480014.1 | XP_021335689.1 | 41 - 42 | 6860 | 86 | ||
| XM_021480015.1 | XP_021335690.1 | 39 - 40 | 5990 | 86 | ||
| XM_021480016.1 | XP_021335691.1 | 36 - 37 | 5772 | 86 | ||
| GenBank mRNA | BC055667.1 | - | 11 - 12 | 1555 | 86 | |
| DQ227765.1 | - | 36 - 37 | 5540 | 86 | ||
More Information
Set Membership: |
|

svila
Genomic Map