Assay Details
| Gene Symbol: svila | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035261.2 | NP_001030338.2 | 37 - 38 | 5552 | 75 | |
| XM_021480011.1 | XP_021335686.1 | 43 - 44 | 7047 | 75 | ||
| XM_021480012.1 | XP_021335687.1 | 42 - 43 | 6977 | 75 | ||
| XM_021480013.1 | XP_021335688.1 | 42 - 43 | 6971 | 75 | ||
| XM_021480014.1 | XP_021335689.1 | 42 - 43 | 6878 | 75 | ||
| XM_021480015.1 | XP_021335690.1 | 40 - 41 | 6008 | 75 | ||
| XM_021480016.1 | XP_021335691.1 | 37 - 38 | 5790 | 75 | ||
| GenBank mRNA | BC055667.1 | - | 12 - 13 | 1573 | 75 | |
| DQ227765.1 | - | 37 - 38 | 5558 | 75 | ||
More Information
Set Membership: |
|

svila
Genomic Map