Assay Details
Gene Symbol: ppp2r2ca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037394.1 | NP_001032471.1 | 8 - 9 | 1187 | 144 | |
XM_021478641.1 | XP_021334316.1 | 7 - 8 | 1246 | 144 | ||
GenBank mRNA | BC109406.1 | - | 8 - 9 | 1187 | 144 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
