Assay Details
Gene Symbol: reep3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037575.1 | NP_001032664.1 | 5 - 6 | 674 | 86 | |
GenBank mRNA | BC110095.1 | - | 5 - 6 | 674 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
