Assay Details
Gene Symbol: pstpip1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037579.1 | NP_001032668.1 | 12 - 13 | 1008 | 84 | |
GenBank mRNA | BC110107.1 | - | 12 - 13 | 1008 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
