Assay Details
| Gene Symbol: pstpip1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037579.1 | NP_001032668.1 | 7 - 8 | 623 | 61 | |
| GenBank mRNA | BC110107.1 | - | 7 - 8 | 623 | 61 | |
More Information
Set Membership: |
|

pstpip1b
Genomic Map