Assay Details
| Gene Symbol: pmp22b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040355.2 | NP_001035445.1 | 4 - 5 | 430 | 60 | |
| GenBank mRNA | BC115330.1 | - | 4 - 5 | 396 | 60 | |
| BC154259.1 | - | 4 - 5 | 415 | 60 | ||
More Information
Set Membership: |
|

pmp22b
Genomic Map