Assay Details
| Gene Symbol: abcg2d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042772.1 | NP_001036237.1 | 10 - 11 | 1430 | 81 | |
| XM_009294223.3 | XP_009292498.3 | 12 - 13 | 1858 | 81 | ||
| GenBank mRNA | DQ223861.1 | - | 10 - 11 | 1430 | 81 | |
More Information
Set Membership: |
|

abcg2d
Genomic Map