Assay Details
Gene Symbol: abcg2d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042772.1 | NP_001036237.1 | 4 - 5 | 539 | 88 | |
XM_009294223.3 | XP_009292498.3 | 5 - 6 | 895 | 88 | ||
GenBank mRNA | DQ223861.1 | - | 4 - 5 | 539 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
