Assay Details
| Gene Symbol: atp2b1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044757.1 | NP_001038222.1 | 3 - 4 | 683 | 103 | |
| XM_009300256.3 | XP_009298531.1 | 3 - 4 | 1258 | 103 | ||
| XM_021474964.1 | XP_021330639.1 | 3 - 4 | 764 | 103 | ||
| XM_021474965.1 | XP_021330640.1 | 3 - 4 | 764 | 103 | ||
| XM_021474966.1 | XP_021330641.1 | 2 - 3 | 418 | 103 | ||
| GenBank mRNA | BC076309.1 | - | 3 - 4 | 103 | ||
| BC092356.1 | - | 3 - 4 | 103 | |||
| BC115143.1 | - | 3 - 4 | 707 | 103 | ||
| BC121780.1 | - | 3 - 4 | 697 | 103 | ||
| BC129304.1 | - | 3 - 4 | 697 | 103 | ||
| BC171622.1 | - | 2 - 3 | 520 | 103 | ||
| DQ242508.1 | - | 3 - 4 | 681 | 103 | ||
| HM449162.1 | - | 2 - 3 | 397 | 103 | ||
More Information
Set Membership: |
|

atp2b1a
Genomic Map