Assay Details
| Gene Symbol: atp2b1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044757.1 | NP_001038222.1 | 4 - 5 | 937 | 89 | |
| XM_009300256.3 | XP_009298531.1 | 4 - 5 | 1512 | 89 | ||
| XM_021474964.1 | XP_021330639.1 | 4 - 5 | 1018 | 89 | ||
| XM_021474965.1 | XP_021330640.1 | 4 - 5 | 1018 | 89 | ||
| XM_021474966.1 | XP_021330641.1 | 3 - 4 | 672 | 89 | ||
| GenBank mRNA | BC092356.1 | - | 4 - 5 | 953 | 89 | |
| BC115143.1 | - | 4 - 5 | 961 | 89 | ||
| BC121780.1 | - | 4 - 5 | 951 | 89 | ||
| BC129304.1 | - | 4 - 5 | 951 | 89 | ||
| BC171622.1 | - | 3 - 4 | 774 | 89 | ||
| DQ242508.1 | - | 4 - 5 | 935 | 89 | ||
| HM449162.1 | - | 3 - 4 | 651 | 89 | ||
More Information
Set Membership: |
|

atp2b1a
Genomic Map