Assay Details
| Gene Symbol: mfge8a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014299.2 | NP_001014321.2 | 1 - 2 | 483 | 66 | |
| GenBank mRNA | BC139634.1 | - | 1 - 2 | 483 | 66 | |
| EH456613.1 | - | 1 - 2 | 126 | 66 | ||
More Information
Set Membership: |
|

mfge8a
Genomic Map