Assay Details
Gene Symbol: b3galt6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045225.1 | NP_001038690.1 | 1 | 449 | 93 | |
XM_005155657.4 | XP_005155714.1 | 2 | 786 | 93 | ||
GenBank mRNA | AM235756.1 | - | 1 | 449 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
