Assay Details
| Gene Symbol: zc3hc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077378.1 | NP_001070846.1 | 1 - 2 | 199 | 59 | |
| GenBank mRNA | BC124810.1 | - | 1 - 2 | 199 | 59 | |
More Information
Set Membership: |
|

zc3hc1
Genomic Map