Assay Details
| Gene Symbol: ano10b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077382.1 | NP_001070850.1 | 12 - 13 | 2006 | 146 | |
| XM_021470507.1 | XP_021326182.1 | 13 - 14 | 2265 | 146 | ||
| XM_021470508.1 | XP_021326183.1 | 12 - 13 | 2125 | 146 | ||
| GenBank mRNA | BC124774.1 | - | 12 - 13 | 2006 | 146 | |
| BC165242.1 | - | 10 - 11 | 1936 | 146 | ||
More Information
Set Membership: |
|

ano10b
Genomic Map