Assay Details
| Gene Symbol: ano10b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077382.1 | NP_001070850.1 | 3 - 4 | 308 | 64 | |
| XM_021470507.1 | XP_021326182.1 | 4 - 5 | 567 | 64 | ||
| XM_021470508.1 | XP_021326183.1 | 3 - 4 | 427 | 64 | ||
| GenBank mRNA | BC124774.1 | - | 3 - 4 | 308 | 64 | |
| BC165242.1 | - | 1 - 2 | 238 | 64 | ||
More Information
Set Membership: |
|

ano10b
Genomic Map