Assay Details
Gene Symbol: glyctk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077388.2 | NP_001070856.2 | 4 - 5 | 892 | 93 | |
NM_001316959.1 | NP_001303888.1 | 4 - 5 | 785 | 93 | ||
XM_005166060.4 | XP_005166117.1 | 4 - 5 | 854 | 93 | ||
XM_005166062.4 | XP_005166119.1 | 4 - 5 | 899 | 93 | ||
XM_021476919.1 | XP_021332594.1 | 5 - 6 | 982 | 93 | ||
GenBank mRNA | BC124662.1 | - | 4 - 5 | 93 | ||
EH584158.1 | - | 4 - 5 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
