Assay Details
Gene Symbol: ppm1la | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077600.1 | NP_001071068.1 | 1 - 2 | 788 | 74 | |
GenBank mRNA | BC125953.1 | - | 1 - 2 | 788 | 74 | |
BC164403.1 | - | 1 - 2 | 434 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
