Assay Details
| Gene Symbol: plod1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077742.2 | NP_001071210.2 | 6 - 7 | 767 | 71 | |
| GenBank mRNA | BC125830.1 | - | 6 - 7 | 767 | 71 | |
| DQ020093.1 | - | 6 - 7 | 705 | 71 | ||
More Information
Set Membership: |
|

plod1a
Genomic Map