Assay Details
| Gene Symbol: cacnb2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081748.2 | NP_001075217.2 | 1250 | 72 | ||
| NM_001166324.1 | NP_001159796.1 | 1008 | 72 | |||
| GenBank mRNA | BC160620.1 | - | 11 - 12 | 908 | 72 | |
| BC162962.1 | - | 11 - 12 | 875 | 72 | ||
| BC162994.1 | - | 11 - 12 | 875 | 72 | ||
| DQ372944.1 | - | 11 - 12 | 908 | 72 | ||
| EU301442.1 | - | 1250 | 72 | |||
| EU301443.1 | - | 1008 | 72 | |||
More Information
Set Membership: |
|

cacnb2b
Genomic Map