Assay Details
| Gene Symbol: cacnb2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC160620.1 | - | 14 - 15 | 1203 | 67 | |
| BC162962.1 | - | 14 - 15 | 1170 | 67 | ||
| BC162994.1 | - | 14 - 15 | 1170 | 67 | ||
| DQ372944.1 | - | 14 - 15 | 1203 | 67 | ||
More Information
Set Membership: |
|

cacnb2b
Genomic Map