Assay Details
| Gene Symbol: eif4ba | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099237.2 | NP_001092707.2 | 4 - 5 | 529 | 98 | |
| XM_005161956.4 | XP_005162013.1 | 4 - 5 | 523 | 98 | ||
| XM_005161957.4 | XP_005162014.1 | 4 - 5 | 523 | 98 | ||
| GenBank mRNA | BC070016.1 | - | 3 - 4 | 472 | 98 | |
| BC129148.1 | - | 3 - 4 | 472 | 98 | ||
| BC142793.1 | - | 4 - 5 | 529 | 98 | ||
More Information
Set Membership: |
|

eif4ba
Genomic Map