Assay Details
| Gene Symbol: bcl6ab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100074.1 | NP_001093544.1 | 3 - 4 | 459 | 64 | |
| XM_005171137.3 | XP_005171194.1 | 3 - 4 | 478 | 64 | ||
More Information
Set Membership: |
|

bcl6ab
Genomic Map