Assay Details
| Gene Symbol: ldb2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131315.3 | NP_571390.1 | 2 - 3 | 369 | 84 | |
| XM_009307714.3 | XP_009305989.1 | 2 - 3 | 396 | 84 | ||
| XM_009307716.3 | XP_009305991.1 | 2 - 3 | 396 | 84 | ||
| GenBank mRNA | AF031377.1 | - | 2 - 3 | 354 | 84 | |
| BC090063.1 | - | 2 - 3 | 369 | 84 | ||
| BC164481.1 | - | 2 - 3 | 264 | 84 | ||
More Information
Set Membership: |
|

ldb2b
Genomic Map