Assay Details
| Gene Symbol: smad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131356.2 | NP_571431.2 | 2 - 3 | 675 | 73 | |
| GenBank mRNA | AF167985.1 | - | 1 - 2 | 407 | 73 | |
| AF174434.1 | - | 1 - 2 | 407 | 73 | ||
| AY309268.1 | - | 2 - 3 | 640 | 73 | ||
| BC086962.1 | - | 2 - 3 | 606 | 73 | ||
More Information
Set Membership: |
|

smad1
Genomic Map