Assay Details
Gene Symbol: appa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131564.3 | NP_571639.1 | 12 - 13 | 2007 | 67 | |
GenBank mRNA | AJ315636.1 | - | 7 - 8 | 1083 | 67 | |
AJ315637.1 | - | 5 - 6 | 739 | 67 | ||
BC068375.1 | - | 12 - 13 | 1888 | 67 | ||
CU638811.1 | - | 11 - 12 | 1714 | 67 | ||
EH435990.1 | - | 4 - 5 | 565 | 67 | ||
JQ994487.1 | - | 11 - 12 | 1721 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
