Assay Details
Gene Symbol: hnf1ba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131880.3 | NP_571955.3 | 5 - 6 | 1361 | 124 | |
XM_021481477.1 | XP_021337152.1 | 5 - 6 | 1366 | 124 | ||
XM_021481478.1 | XP_021337153.1 | 5 - 6 | 1070 | 124 | ||
GenBank mRNA | AF244140.2 | - | 5 - 6 | 1304 | 124 | |
AF430840.1 | - | 5 - 6 | 1368 | 124 | ||
AY336602.1 | - | 5 - 6 | 1275 | 124 | ||
AY569309.1 | - | 5 - 6 | 1328 | 124 | ||
BC129147.1 | - | 5 - 6 | 1336 | 124 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
