Assay Details
| Gene Symbol: hnf1ba | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131880.3 | NP_571955.3 | 8 - 9 | 1787 | 93 | |
| XM_021481477.1 | XP_021337152.1 | 7 - 8 | 1873 | 93 | ||
| XM_021481478.1 | XP_021337153.1 | 7 - 8 | 1577 | 93 | ||
| GenBank mRNA | AF244140.2 | - | 8 - 9 | 1730 | 93 | |
| AF430840.1 | - | 8 - 9 | 1794 | 93 | ||
| AY336602.1 | - | 8 - 9 | 1701 | 93 | ||
| AY569309.1 | - | 8 - 9 | 1754 | 93 | ||
| BC129147.1 | - | 8 - 9 | 1762 | 93 | ||
More Information
Set Membership: |
|

hnf1ba
Genomic Map