Assay Details
| Gene Symbol: naca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193352.1 | NP_001180281.1 | 5 - 6 | 2784 | 69 | |
| NM_173264.2 | NP_775371.1 | 4 - 5 | 339 | 69 | ||
| GenBank mRNA | AY099510.1 | - | 4 - 5 | 275 | 69 | |
| AY398384.1 | - | 4 - 5 | 279 | 69 | ||
| BC122432.1 | - | 4 - 5 | 324 | 69 | ||
| BC164312.1 | - | 3 - 4 | 267 | 69 | ||
More Information
Set Membership: |
|

naca
Genomic Map