Assay Details
Gene Symbol: rock2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174863.2 | NP_777288.1 | 25 - 26 | 3516 | 70 | |
GenBank mRNA | AF295804.1 | - | 25 - 26 | 3516 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
