Assay Details
| Gene Symbol: timp2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_182874.1 | NP_878294.1 | 3 - 4 | 564 | 93 | |
| GenBank mRNA | AY324194.1 | - | 3 - 4 | 564 | 93 | |
| BC115244.1 | - | 3 - 4 | 509 | 93 | ||
| BC125891.1 | - | 3 - 4 | 561 | 93 | ||
| BC165045.1 | - | 3 - 4 | 378 | 93 | ||
More Information
Set Membership: |
|

timp2a
Genomic Map