Assay Details
| Gene Symbol: birc5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_194397.3 | NP_919378.1 | 2 - 3 | 352 | 73 | |
| GenBank mRNA | AY057057.1 | - | 2 - 3 | 270 | 73 | |
| BC056739.1 | - | 2 - 3 | 345 | 73 | ||
| EH547926.1 | - | 2 - 3 | 377 | 73 | ||
| EH589060.1 | - | 2 - 3 | 342 | 73 | ||
More Information
Set Membership: |
|

birc5a
Genomic Map