Assay Details
| Gene Symbol: egfra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_194424.1 | NP_919405.1 | 2 - 3 | 395 | 105 | |
| GenBank mRNA | AY154658.1 | - | 2 - 3 | 395 | 105 | |
| AY332223.1 | - | 2 - 3 | 300 | 105 | ||
| AY533377.1 | - | 2 - 3 | 300 | 105 | ||
| AY533378.1 | - | 2 - 3 | 300 | 105 | ||
| AY533379.1 | - | 2 - 3 | 300 | 105 | ||
| BC163725.1 | - | 2 - 3 | 266 | 105 | ||
| KT023304.1 | - | 2 - 3 | 231 | 105 | ||
More Information
Set Membership: |
|

egfra
Genomic Map