Assay Details
Gene Symbol: wbp2nl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199710.1 | NP_956004.1 | 2 - 3 | 279 | 72 | |
GenBank mRNA | BC053232.1 | - | 2 - 3 | 279 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
