Assay Details
| Gene Symbol: mapre1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282033.1 | NP_001268962.1 | 3 - 4 | 442 | 102 | |
| NM_199864.2 | NP_956158.2 | 3 - 4 | 442 | 102 | ||
| GenBank mRNA | BC044167.1 | - | 3 - 4 | 394 | 102 | |
| BC065871.1 | - | 3 - 4 | 425 | 102 | ||
| BC165535.1 | - | 2 - 3 | 295 | 102 | ||
| EH547703.1 | - | 3 - 4 | 424 | 102 | ||
More Information
Set Membership: |
|

mapre1a
Genomic Map