Assay Details
| Gene Symbol: mapre1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199864.2 | NP_956158.2 | 4 - 5 | 648 | 85 | |
| GenBank mRNA | BC044167.1 | - | 4 - 5 | 600 | 85 | |
| BC165535.1 | - | 3 - 4 | 501 | 85 | ||
| EH547703.1 | - | 4 - 5 | 630 | 85 | ||
More Information
Set Membership: |
|

mapre1a
Genomic Map