Assay Details
| Gene Symbol: atp6v1g1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199934.1 | NP_956228.1 | 2 - 3 | 244 | 90 | |
| GenBank mRNA | AF210636.1 | - | 1 - 2 | 75 | 90 | |
| BC059611.1 | - | 2 - 3 | 244 | 90 | ||
| BC152208.1 | - | 2 - 3 | 263 | 90 | ||
More Information
Set Membership: |
|

atp6v1g1
Genomic Map