Assay Details
| Gene Symbol: prrx1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200050.1 | NP_956344.1 | 2 - 3 | 787 | 67 | |
| XM_005160643.4 | XP_005160700.1 | 2 - 3 | 993 | 67 | ||
| XM_017352441.2 | XP_017207930.1 | 2 - 3 | 993 | 67 | ||
| GenBank mRNA | BC064671.1 | - | 2 - 3 | 787 | 67 | |
More Information
Set Membership: |
|

prrx1b
Genomic Map