Assay Details
| Gene Symbol: maff | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200336.1 | NP_956630.1 | 2 - 3 | 223 | 64 | |
| GenBank mRNA | AB167543.1 | - | 2 - 3 | 218 | 64 | |
| BC053121.1 | - | 2 - 3 | 223 | 64 | ||
More Information
Set Membership: |
|

maff
Genomic Map