Assay Details
| Gene Symbol: riox2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200337.1 | NP_956631.1 | 3 - 4 | 641 | 94 | |
| XM_021479115.1 | XP_021334790.1 | 4 - 5 | 720 | 94 | ||
| XM_021479116.1 | XP_021334791.1 | 4 - 5 | 717 | 94 | ||
| XM_021479117.1 | XP_021334792.1 | 4 - 5 | 720 | 94 | ||
| GenBank mRNA | BC053125.1 | - | 3 - 4 | 641 | 94 | |
More Information
Set Membership: |
|

riox2
Genomic Map