Assay Details
| Gene Symbol: fgfrl1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200376.1 | NP_956670.1 | 6 - 7 | 1146 | 159 | |
| GenBank mRNA | AJ574916.1 | - | 6 - 7 | 1147 | 159 | |
| BC053245.1 | - | 6 - 7 | 1146 | 159 | ||
| CU458922.1 | - | 6 - 7 | 1147 | 159 | ||
More Information
Set Membership: |
|

fgfrl1a
Genomic Map