Assay Details
| Gene Symbol: nr2f1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200592.1 | NP_956886.1 | 2 - 3 | 1183 | 63 | |
| XM_021479121.1 | XP_021334796.1 | 2 - 4 | 1231 | 63 | ||
| GenBank mRNA | BC056748.1 | - | 2 - 3 | 1183 | 63 | |
| BC065651.1 | - | 2 - 4 | 1179 | 63 | ||
More Information
Set Membership: |
|

nr2f1b
Genomic Map