Assay Details
Gene Symbol: adam8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200637.1 | NP_956931.1 | 18 - 19 | 1987 | 94 | |
GenBank mRNA | BC057428.1 | - | 18 - 19 | 1987 | 94 | |
BC165674.1 | - | 18 - 19 | 1985 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
