Assay Details
| Gene Symbol: adam8a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200637.1 | NP_956931.1 | 19 - 20 | 2099 | 83 | |
| GenBank mRNA | BC057428.1 | - | 19 - 20 | 2099 | 83 | |
| BC165674.1 | - | 19 - 20 | 2097 | 83 | ||
More Information
Set Membership: |
|

adam8a
Genomic Map