Assay Details
Gene Symbol: mgat1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200676.1 | NP_956970.1 | 1 - 2 | 704 | 62 | |
XM_005157757.3 | XP_005157814.1 | 2 - 3 | 1170 | 62 | ||
GenBank mRNA | BC058297.1 | - | 1 - 2 | 704 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
